Key Documents
Z421502
Sigma-Aldrich
Pipe cap, for Kilo-Lab® system
stainless steel, NPTF 1/4 in.
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Pricing and availability is not currently available.
material
stainless steel
NPTF
1/4 in.
General description
For capping end of transfer line having 1/4 in. MNPT opening.
Legal Information
Kilo-Lab is a registered trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)