Z420093
Scienceware® Vikem®-coated C-shaped lead ring
fits flask, 125 ‑ 500 mL
Select a Size
$53.50
Select a Size
About This Item
$53.50
Recommended Products
material
lead (Vikem vinyl coated)
sterility
non-sterile
feature
fits flask 125 ‑ 500 mL
packaging
pack of 1
manufacturer/tradename
Bel-Art F18308-0500
weight
230 g
Looking for similar products? Visit Product Comparison Guide
General description
- Add to fluid handling setups already in place
- Accommodate wider vessels by pulling on end of rings to expand I.D.
- Rings fit neatly around the neck of filtering flasks with side-arm tubing connections
- Green in color
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service