Z409251
Aldrich® Schlaker reaction tube
capacity 50 mL, plain joint
Select a Size
$343.00
Estimated to ship onJune 01, 2025
Select a Size
About This Item
$343.00
Estimated to ship onJune 01, 2025
Recommended Products
material
colorless tube
glass tube
round bottom
sterility
non-sterile
feature
graduations: no
neck thread PUSH FIT (14/20 top joint)
capacity
50 mL
binding type
non-treated surface
Related Categories
General description
Made of heavy-wall glass, with glass stopcock and frit (porosity 25-50 mm) on side-arm, and a plain or a threaded 14/20 top joint. Compatible with Schlenk glassware. Compatible with the Aldrich All-in-One Glassware Kit.
Schlaker Tube Technique
1. Place resin in the tube
2. Evacuate and purge with N2 to remove oxygen via a rubber septum or a septum-inlet adapter in the top joint
3. Stir gently while heating in an oil bath for high temperatures, or place on a shaker for room temperature reactions
4. When the reaction is complete, apply positive N2 pressure through the septum in the top joint, and vacuum to the side-arm
5. Tilt the Schlaker tube on its side to allow solvent and excess reagents to flow out, leaving the resin behind
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)