Z405671
J. Young general-purpose stopcock
straight through, PTFE, side arm O.D. 8 mm
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
material
PTFE
manufacturer/tradename
J. Young
side arm O.D.
8 mm
Looking for similar products? Visit Product Comparison Guide
General description
- Grease-free PTFE stopcock
- Temp. range: -18 to 60°C
- 0 to 5 mm flow cross-section
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service