No, the battery is not replaceable.
Z314951
Hirschmann® pipetus® rechargeable pipette controller
AC/DC input 100 - 240 V AC, supplied with charger and a US, UK and flat 2-pin Europlug
Synonym(s):
Pipette Controller
About This Item
Recommended Products
material
plastic
silicone
feature
CE compliant
manufacturer/tradename
Hirschmann 9907200
volume
0.1-200 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Rechargeable battery, up to 50 hour use, with charge indicator
- Use with volumetric, serological and plastic pipettes, volumes 0.1 to 200 mL
- Display of selected pipetting mode, EX or blow-out
- Indication of the selected pipetting speed
- Buzzer signal sounds when battery capacity reaches below 10%
- Adjustable suction speed, depending upon volume and viscosity of medium
- One button operation
- No trailing wires
- Fine dosing valve
- Silicone adapter with exchangeable, internal filter, holds all types of pipettes firm and tight
- Charges automatically just by inserting into stand
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
is it possible to have the battery replaced? If so how much would that cost?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service