Z309222
Quickfit® Allihn condenser
L 250 mm, joint: ST/NS 35/35
Synonym(s):
CX7/05/SC
All Photos(1)
About This Item
UNSPSC Code:
41121800
Joint
joint: ST/NS35/35
manufacturer/tradename
Quickfit CX 7/05/SC
availability
not available in USA and Canada
L
250 mm
surface area
130 cm2
General description
Bulb sections give greater surface area for more cooling than the corresponding Liebig type condenser. Generally used for reflux. Condensers have plain stem tops.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)