Key Documents
Z304824
Quickfit® drying tube
joint: ST/NS 14/23
Synonym(s):
MF23/1
Select a Size
1 EA
$66.20
$66.20
Estimated to ship onApril 14, 2025
Select a Size
Change View
1 EA
$66.20
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$66.20
Estimated to ship onApril 14, 2025
material
colorless tube
glass tube
Joint
joint: ST/NS14/23
sterility
non-sterile
feature
graduations: no
neck thread PUSH FIT (14/23 cone size)
manufacturer/tradename
Quickfit MF 23/1
availability
not available in USA and Canada
binding type
non-treated surface
General description
Angled. Retain desiccant with plug of glass wool.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)