Key Documents
Z303917
Quickfit® flat flange lids for reaction flasks
center joint: ST/NS 34/35, parallel socket joint: 19/26, 10° side socket Joints: 19/26 (2)
Synonym(s):
MAF5/2
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
material
borosilicate glass
label
B
Joint
10° side socket Joints: 19/26 (2)
15° side socket joint
5° side socket joint
center joint: ST/NS34/35
parallel socket joint: 19/26
sterility
non-sterile
packaging
pack of 1
manufacturer/tradename
Quickfit MAF 5/2
availability
not available in USA and Canada
General description
Single and multi-socket lids for reaction flasks for simple extraction, reflux or for the introduction of a variety of stirrers, dropping funnels, temperature probes etc.
Use clip to secure lid to flasks, purchase separately.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)