Key Documents
Z301159
Quickfit® expansion adapter
top female joint: ST/NS 34/35, bottom male joint: ST/NS 19/26
Synonym(s):
XA52
Select a Size
1 EA
$93.80
$93.80
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$93.80
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$93.80
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
female joint: ST/NS top34/35
male joint: ST/NS bottom19/26
manufacturer/tradename
Quickfit XA 52
availability
not available in USA and Canada
not available in USA and Canada
General description
These adapters connect items with larger cone to a piece of glassware with a smaller size socket.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)