Key Documents
Z301078
Quickfit® Reduction Adapters
top female joint: ST/NS 34/35, bottom male joint: ST/NS 50/42
Synonym(s):
DA58
Select a Size
1 EA
$70.90
$70.90
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$70.90
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$70.90
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
female joint: ST/NS top34/35
male joint: ST/NS bottom50/42
manufacturer/tradename
Quickfit DA 58
availability
not available in USA and Canada
General description
These adapters connect items with a smaller cone to a piece of glassware with a larger size socket.
Legal Information
Quickfit is a registered trademark of SciLabware Limited
Choose from one of the most recent versions:
Certificates of Analysis (COA)