Key Documents
Z282464
Spinpak® stir bar
size 1 1/2 in. × 5/16 in.
Select a Size
1 EA
$42.90
$42.90
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$42.90
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
$42.90
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
octagonal PTFE-coated
packaging
pack of 6 ea
manufacturer/tradename
Bel-Art F37157-0015
size
1 1/2 in. × 5/16 in.
General description
The four best selling Spinbar® sizes in a convenient blister pack. Each pack contains octagonal magnetic Spinbars of one size.
Legal Information
Spinbar is a registered trademark of SP Scienceware, a division of SP
Spinpak is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)