Key Documents
Z282073
Coors alumina pestle
for Z282022
Select a Size
1 EA
$124.00
$124.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
Select a Size
Change View
1 EA
$124.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
$124.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
material
alumina
packaging
pack of 1 ea
manufacturer/tradename
Coorstek 60365
pestle L
74 mm
Related Categories
General description
99.5% alumina. Extremely hard and completely nonabsorbent surface. Order mortar separately.
Legal Information
Coors is a trademark of Coorstek, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)