Key Documents
Z278351
ACE Micro/Mini-Lab® Hickman-Hinkle stillhead
joint: ST/NS 14/10 top and bottom, head size 10 mL
Select a Size
1 EA
$124.00
$124.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$124.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
$124.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS14/10 top and bottom
manufacturer/tradename
Ace Glass 959962
head size
10 mL
Related Categories
General description
For conducting simple and fractional distillations. Has threaded side port with septum and cap for syringe access to well contents and an elongated section below well to accommodate the PTFE spinning band. 10 mL still has 14/10 threaded outer joint at top and 14/10 inner joint at bottom. Top outer joint supplied with cap, septum, and O-ring.
Legal Information
Micro/Mini-Lab is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)