Skip to Content
Merck
All Photos(1)

Key Documents

Z275409

Sigma-Aldrich

Ostwald viscometer tube

outflow time of 80 to 100s with water at 20°C

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$787.00

$787.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025



Select a Size

Change View
1 EA
$787.00

About This Item

UNSPSC Code:
41111700
NACRES:
PB.36

$787.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


manufacturer/tradename

Ace Glass 798510

Related Categories

General description

For the determination of relative viscosities of liquids. Adjusted for outflow time of 80 to 100s with water at 20°C.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–3 of 3 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. Does Ostwald viscometer, Product Z275409, need to be calibrated prior to use?

    1 answer
    1. We do recommend that you calibrate the instrument prior to use, which can be done by using materials with known viscosities.

      Helpful?

  3. Do you have instructions for using the Ostwald viscometer, Product Z275409?

    1 answer
    1. We do have instructions for using this viscometer. Please call your local Technical Service, and we can send you these instructions.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service