Skip to Content
Merck
All Photos(1)

Key Documents

Z273287

Sigma-Aldrich

Terg-a-zyme® enzyme detergent

pack of 11 kg

Synonym(s):

tergazyme

Sign Into View Organizational & Contract Pricing

Select a Size

11 KG
$330.00

$330.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
11 KG
$330.00

About This Item

UNSPSC Code:
47131800
NACRES:
NB.35

$330.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

packaging

pack of 11 kg

manufacturer/tradename

Alconox, Inc. 1325 (4 x 3.8 L)

General description

Tergazyme® is a combination of ALCONOX® and protease enzyme for presoaking proteinaceous soils and hard-to-remove stains.

Used to clean: Hospital instruments, dairy equipment, laboratory ware, reverse osmosis and ultrafiltration membranes and units, sampling apparatus, pharmaceutical apparatus, cosmetics manufacturing equipment, tubing, pipes, optical parts, process equipment, industrial parts, desalination plants, tanks and reactors.Authorized by USDA for use in federally inspected meat and poultry plants. Passes inhibitory residue test for water analysis. FDA certified.

Used to remove: Soil, grit, grime, blood, tissue, grease, fats, oils. Aroteinaceous soils, dairy proteins, particulates, solvents and bioreactor residue.

Surfaces cleaned: Corrosion inhibited formulation recommended for glass, metal, stainless steel, porcelain, ceramic, plastic, rubber and fiberglass. Can be used on soft metals such as copper, aluminum, zinc and magnesium if rinsed promptly. Corrosion testing may be advisable.

Cleaning method: Soak, brush, sponge, cloth, ultrasonic, flow through clean-in-place. Will foam—not for spray or machine use.

Directions: Make a fresh 1% solution (2 1/2 tbsp per gal, 1 1/4 oz per gal, or 10 g per L) in cold or warm water. If available, use warm water below 130 °F (55 °C). Clean by soak, circulate, wipe, or ultrasonic method. Follow manufacturer′s directions for filter membrane cleaning.
Not for spray machines: will foam. RINSE THOROUGHLY—preferably with running water. For critical cleaning, do final or all rinsing in distilled, deionized, or purified water. For food contact surfaces, rinse with potable water.

Features and Benefits

Tergazyme® enzyme detergent:
  • Concentrated to save you money
  • Biodegradable and readily disposable
  • Replaces corrosive acids and hazardous solvents
  • Protease enzyme removes proteinaceous soils, tissue, blood and body fluids
  • Free rinsing to provide reliable results and no interfering residues
  • Use to pass cleaning validation tests for lab accreditation and plant inspection approval

Legal Information

ALCONOX is a registered trademark of Alconox, Inc.
Tergazyme is a registered trademark of Alconox, Inc.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nathan Moore et al.
Scientific reports, 9(1), 15101-15101 (2019-10-24)
The development and approval of engineered cellular therapies are revolutionizing approaches to treatment of diseases. However, these life-saving therapies require extensive use of inefficient bioprocessing equipment and specialized reagents that can drive up the price of treatment. Integration of new
Manuel S Brugger et al.
Biosensors & bioelectronics, 173, 112807-112807 (2020-11-23)
A Love-wave based biosensor is introduced for analyzing a standardized wound healing assay by observing cell growth and quantifying cell detachment processes. Utilizing the piezoelectric material LiTaO3 36° XY-cut with a thin SiO2-cover layer, shear horizontal surface acoustic waves (SAW)
Michele Dollt et al.
Frontiers in neuroscience, 14, 552876-552876 (2020-10-20)
The performance of electrode arrays insulated by low-temperature atomic layer deposited (ALD) titanium dioxide (TiO2) or hafnium dioxide (HfO2) for culture of electrogenic cells and for recording of extracellular action potentials is investigated. If successful, such insulation may be considered
Larissa Höfling et al.
Scientific reports, 10(1), 5248-5248 (2020-04-07)
Retinal implants are used to replace lost photoreceptors in blind patients suffering from retinopathies such as retinitis pigmentosa. Patients wearing implants regain some rudimentary visual function. However, it is severely limited compared to normal vision because non-physiological stimulation strategies fail

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service