Z266175
Nalgene® FEP bottle
capacity 2000 mL, Narrow-mouth, clear Bottle
Synonym(s):
fep bottles, nalgene bottles
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
Recommended Products
material
ETFE cap
round translucent FEP bottle
Joint
Joints threaded neck (38)
feature
with cap
packaging
pack of 1 ea
manufacturer/tradename
Nalgene® 1600-0064
bottle diam. × H
121 mm × 245 mm
capacity
2000 mL
color
clear Bottle
white Screw closure
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Perform trace metal analysis and organic solvent applications with temperature and chemical resistant Nalgene®PTFEtef® FEP Wide Mouth Bottles. Made entirely of fluoropolymer, these bottles are autoclavable and can be ultra-cleaned in boiling nitric acid.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Teflon is a registered trademark of E. I. du Pont de Nemours and Company
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service