Skip to Content
Merck
All Photos(1)

Key Documents

Z265918

Sigma-Aldrich

Microporous carbon boiling chips

weight 227 g

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$48.10

$48.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EA
$48.10

About This Item

EC Number:
UNSPSC Code:
41104812
NACRES:
NB.43

$48.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

packaging

pack of 1 ea

weight

227 g

Related Categories

General description

Eliminates bumping of liquids in boiling operations and distillations.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service