Key Documents
Z251860
Supelco
Michel-Miller column stopper
Ace-Thred for #15
Select a Size
1 EA
$118.00
$118.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$118.00
About This Item
UNSPSC Code:
12000000
NACRES:
SB.52
$118.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Request a Bulk Order
manufacturer/tradename
Ace Glass 580311
technique(s)
LPLC: suitable
fitting
Ace-Thred for #15
General description
PTFE for use at either end of any chromatography column with Ace-Threds. Makes a leak-tight seal without O-rings.
Choose from one of the most recent versions:
Certificates of Analysis (COA)