Z250031
Wheaton narrow mouth bottles with caps
capacity 8 oz (250 mL), amber, white polypropylene cap, PTFE flat liner
Synonym(s):
Wheaton bottle, Wheaton bottles, narrow mouth
Select a Size
$164.00
Select a Size
About This Item
$164.00
Recommended Products
material
PTFE flat liner
amber bottle
glass bottle
round bottle
white polypropylene cap
Joint
Joints threaded neck (24-400)
feature
with cap
packaging
pkg of 12 ea
manufacturer/tradename
WHEATON® W216845
capacity
8 oz (250 mL)
diam. × H
60 mm × 136 mm
color
amber
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Choice of cap styles:
- White PP cap with PTFE or polyvinyl liner
- Black plastic cap with Poly-Seal® PE cone liner
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service