Z248010
Syringe PP/PE without needle
luer lock tip, centered, capacity 5 mL, graduated, 0.2 mL, sterile
Synonym(s):
Luer lock syringes
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
sterility
sterile
feature
graduated 0.2 mL
packaging
pkg of 100 ea individually wrapped
capacity
5 mL
Looking for similar products? Visit Product Comparison Guide
General description
These polypropylene syringes with Luer tips, may be used with any size needle. The rigid polyethylene plungers of these disposable syringes are slightly oversized, creating a tight seal against the walls of the semi-flexible PP barrel, as well as providing an exceptionally smooth draw. No contaminating lubricants added.
Syringes are individually packaged (sterile) except for bulk packed products; these are supplied in plastic bags (non-sterile).
Order needles separately.
Syringes are individually packaged (sterile) except for bulk packed products; these are supplied in plastic bags (non-sterile).
Order needles separately.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Frontiers in bioengineering and biotechnology, 8, 554565-554565 (2020-11-17)
Biomineralized collagen with intrafibrillar calcium phosphate mineral provides an excellent mimic of the composition and structure of the extracellular matrix of bone, from nano- to micro-scale. Scaffolds prepared from this material have the potential to become the next-generation of synthetic
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service