Key Documents
Z247758
Coors high-alumina crucible cover
diam. 59 mm
Select a Size
1 EA
$75.20
$75.20
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$75.20
About This Item
UNSPSC Code:
41122200
NACRES:
NB.43
$75.20
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
packaging
pack of 1 ea
manufacturer/tradename
Coorstek 65613
diam.
59 mm
General description
Covers may not fit exactly on crucibles due to unavoidable wall thickness variations within the crucible.
For Z247642
Legal Information
Coors is a trademark of Coorstek, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)