Key Documents
Z236810
Crew® wipers
L × W 11 1/2 in. × 12 in.
Select a Size
100 EA
$202.00
$202.00
Estimated to ship onApril 15, 2025
Select a Size
Change View
100 EA
$202.00
About This Item
UNSPSC Code:
41103200
NACRES:
NB.35
$202.00
Estimated to ship onApril 15, 2025
packaging
pkg of 100 wipes
manufacturer/tradename
Kimberly Clark
L × W
11 1/2 in. × 12 in.
General description
Low lint wipers for Class 100 assembly operations and other clean room environments. Soft, absorbent, nonwoven fabric eliminates surface fibers and traps particles. Won′t scratch sensitive surfaces and absorbs more than four times its weight in liquids. Acid resistant-won′t break down or disintegrate when exposed to chemicals. Packaged in anti-static, double polybag.
Legal Information
Crew is a registered trademark of Kimberly-Clark Worldwide, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)