Key Documents
Z233463
Sigma-Aldrich
Aldrich® Schlenk-type adapter
Offset with septum-inlet, male joint: ST/NS 14/20
Select a Size
1 EA
$128.00
$128.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
Select a Size
Change View
1 EA
$128.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
$128.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 01, 2025
Joint
female joint ST/NS14/20
male joint: ST/NS14/20
General description
Thermoplastic screw caps with Viton O-rings usable to 140 deg C.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)