Z231835
Duran® beakers
capacity 150 mL, low form
Synonym(s):
211062908
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass 3.3
feature
graduations
manufacturer/tradename
DURAN®
Schott
H × O.D.
80 mm × 60 mm
capacity
150 mL
Looking for similar products? Visit Product Comparison Guide
General description
With spout. Meets ISO 3819 and DIN 12331 standards.
Features and Benefits
• Complies with ISO 3819• Manufactured from borosilicate glass 3.3• Spout for clean pouring• Fired-on, highly durable white ceramic• Easy-to-read scale with graduation marks to estimate the content
Other Notes
Browse all glass beakers
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service