Key Documents
Z223646
Sigma-Aldrich
Aldrich® septum-inlet adapter, female
Screw-thread, joint: ST/NS 29/32
Select a Size
1 EA
$96.30
$96.30
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
Select a Size
Change View
1 EA
$96.30
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$96.30
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
Joint
joint: ST/NS29/32
Related Categories
General description
Septum-inlets require septum Z565695 or Z565679 except where noted.
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)