Skip to Content
Merck
All Photos(1)

Key Documents

Z220469

Sigma-Aldrich

Deoxygenation apparatus

flask size 250 mL, joint: ST/NS 14/20

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,050.00

$1,050.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability


Select a Size

Change View
1 EA
$1,050.00

About This Item

UNSPSC Code:
41100000
NACRES:
NB.43

$1,050.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Joint

joint: ST/NS14/20

flask size

250 mL

General description

One-piece condenser/flask unit for degassing solvents. Flask has angled ST/NS: joint and 7 mm gas dispersion tube (porosity 25-50 μm) with glass stopcock.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service