Key Documents
Z217530
Flexible stirrer shaft coupling
for glass shaft O.D. 10 mm, for motor shaft O.D. 3/8 in.
Select a Size
1 EA
$112.00
$112.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
Select a Size
Change View
1 EA
$112.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.72
$112.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
General description
Flexible coupling allows angular and parallel misalignment of glass stirring shaft without breakage. Neoprene rubber sleeve body absorbs shock and vibration and has great torsional stiffness for quiet, smooth running. PTFE sleeve slides over end of glass shaft. SS set screw secures other end of coupling to motor shaft.
Choose from one of the most recent versions:
Certificates of Analysis (COA)