Key Documents
Z202622
Sigma-Aldrich
Aldrich® Luer lock adapter
male joint: ST/NS 24/40 to male Luer lock
Select a Size
1 EA
$377.00
$377.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$377.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
$377.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
male joint: ST/NS24/40 to male Luer lock
Related Categories
General description
PTFE needle valve and 11/2 in. (38 mm) drip tip.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)