Key Documents
Z202312
Sigma-Aldrich
Pinch clamps for SJ joints
joint: SJ 29/15
Select a Size
1 EA
$71.90
$71.90
Estimated to ship onMay 29, 2025
Select a Size
Change View
1 EA
$71.90
About This Item
UNSPSC Code:
41122413
NACRES:
NB.35
$71.90
Estimated to ship onMay 29, 2025
material
stainless steel
Joint
joint: SJ29/15
General description
Use with flash-chromatography ball jointed assemblies and rotary evaporator receiver flasks.
Choose from one of the most recent versions:
Certificates of Analysis (COA)