Z190373
Aldrich® flask-type sprayer
size 75 mL
Synonym(s):
Chemical Resistant Spray Flask
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
technique(s)
thin layer chromatography (TLC): suitable
size
75 mL
Looking for similar products? Visit Product Comparison Guide
General description
75 mL Erlenmeyer flask with sprayer head (#15 Ace-Thred)
Connect using the barb end to a low-pressure gas or air source of <5 psi. By positioning your thumb over the vent hole you can control the spray pattern.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
replacement part
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service