Merck
All Photos(4)
Key Documents
Z180173
Supelco
Bottom-drip adapter for large columns
Luer tip, 1 mm bore, PTFE
Select a Size
1 EA
$336.00
$336.00
Estimated to ship onApril 15, 2025
Request a Bulk Order
Select a Size
Change View
1 EA
$336.00
About This Item
UNSPSC Code:
12000000
NACRES:
SB.52
$336.00
Estimated to ship onApril 15, 2025
Request a Bulk Order
material
PTFE
manufacturer/tradename
Ace Glass 583761
technique(s)
LPLC: suitable
General description
Shallow taper of upper end minimizes mixing below the PE packing support disc (included). All adapters have an Ace Thred #50 and include one FETFE® O-ring.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)