Z178187
Scienceware® heavy-duty lab-type polypropylene funnel
capacity 30 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
packaging
pack of 6 ea
manufacturer/tradename
Bel-Art F14694-0000
L
38.1 mm
capacity
30 mL
diam.
54.0 mm
stem O.D.
14.3 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Funnels have unique ridges to prevent vapor lock. Recommended for continuous laboratory work.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service