Key Documents
Z178098
Scienceware® heavy-duty test-tube rack
Holds 32 x 16-20 mm tubes
Select a Size
1 EA
$55.70
$55.70
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$55.70
About This Item
UNSPSC Code:
41122804
NACRES:
NB.35
$55.70
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
polypropylene
feature
Holds 32 x 16-20 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F18857-0000
L × W × H
213 mm × 110 mm × 127 mm
General description
Natural PP, stackable. Steam autoclavable to 250°F (121°C). Rows lettered and numbered for easy identification. Base plate has rounded wells with drain holes; end plates with hand grips.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)