Z177415
Scienceware® polypropylene scoops
capacity 50 mL
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
Bel-Art F367480000
bowl size
76 mm × 40 mm × 29 mm
capacity
50 mL
handle size
100 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Molded, natural color.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service