Key Documents
Z177016
Sigma-Aldrich
LDPE wash bottles
capacity 250 mL, Wide-mouth
Synonym(s):
bottle lab, laboratory bottles, wash bottle
Select a Size
12 EA
$86.30
$86.30
Estimated to ship onMay 20, 2025
Select a Size
Change View
12 EA
$86.30
About This Item
UNSPSC Code:
41103209
NACRES:
NB.17
$86.30
Estimated to ship onMay 20, 2025
material
low-density polyethylene bottle
natural bottle
polypropylene cap
wide-mouth bottle
Joint
Joints threaded neck (38)
feature
label no label
vented
with cap
packaging
pack of 12 ea
manufacturer/tradename
Bel-Art F11620-0250
capacity
250 mL
General description
Consist of one-piece cap and nozzle plus delivery tube. Deliver single drops or a needle-sharp stream. Tip may be cut to increase delivery amounts. Polyallomer cap.
Choose from one of the most recent versions:
Certificates of Analysis (COA)