Key Documents
Z173479
Sigma-Aldrich
Double-ended filter with a glass stopcock on each of the two side-arms
capacity 60 mL, male Joints: ST/NS 14/20 (2)
Select a Size
1 EA
$678.00
$678.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$678.00
About This Item
UNSPSC Code:
41104925
NACRES:
NB.43
$678.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
label
BB
Joint
male Joints: ST/NS14/20 (2)
capacity
60 mL
pore size
25-50 μm porosity
Related Categories
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Articles
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.