Key Documents
Z171700
Ace double-jacketed coiled condenser
jacket L 400 mm
Select a Size
1 EA
$602.00
$602.00
Estimated to ship onApril 07, 2025
Select a Size
Change View
1 EA
$602.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$602.00
Estimated to ship onApril 07, 2025
Joint
joint ST/NS24/40
manufacturer/tradename
Ace Glass 595515
jacket L
400 mm
General description
Extra cooling capacity for volatile materials. Compact design retards diffusion.
Choose from one of the most recent versions:
Certificates of Analysis (COA)