Key Documents
Z169013
Sigma-Aldrich
Porcelain berl saddles
pkg of 250 ea
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
packaging
pkg of 250 ea
General description
6 mm. Ideal for vacuum distillation, absorption, and extraction. Surface area: 1.15 m2/m3 In packages of approx. 250 (200 cc).
Maximum temperature: 1100°C.
Choose from one of the most recent versions:
Certificates of Analysis (COA)