Key Documents
Z168130
Ampule breaker/collar
For 11-20 mL ampules
Select a Size
1000 EA
$401.00
$401.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1000 EA
$401.00
About This Item
UNSPSC Code:
41121807
NACRES:
NB.43
$401.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
General description
How to Use:
1. Slip polyethylene breaker/ collar over top of ampule.
2. Grip bottom of ampule with one hand and breaker/collar with other.
3. Gently snap top of ampule off.
4. Discard collar and ampule top.
Choose from one of the most recent versions:
Certificates of Analysis (COA)