Z155616
Griffin-type SS beaker
capacity 3,000 mL, with handle
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
material
stainless steel
feature
graduations: no
H × O.D.
178 mm × 154 mm
capacity
3,000 mL
Looking for similar products? Visit Product Comparison Guide
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service