Z155586
Griffin-type SS beaker
capacity 4,000 mL, without handle
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
material
stainless steel
feature
graduations: no
H × O.D.
225 mm × 156 mm
capacity
4,000 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service