Skip to Content
Merck
All Photos(1)

Key Documents

Z153338

Sigma-Aldrich

NMR tube caps

NMR tube caps

NMR tube size 5 mm

Synonym(s):

cap tubes, nmr tubes, plastic end cap

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$26.00

$26.00


Estimated to ship onMarch 15, 2025



Select a Size

Change View
100 EA
$26.00

About This Item

UNSPSC Code:
41121800
NACRES:
SB.32

$26.00


Estimated to ship onMarch 15, 2025


packaging

pkg of 100 ea

NMR tube size

5 mm

color

blue

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service