Key Documents
Z152439
Sigma-Aldrich
Aldrich® mercury bubbler
joint: ST/NS 24/40, capacity 40 mL
Synonym(s):
bubblers, scientific bubblers
Select a Size
1 EA
$1,100.00
$1,100.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$1,100.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$1,100.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Joint
joint: ST/NS24/40
capacity
40 mL
Related Categories
General description
Permits increased pressure within reaction vessels. Internal reservoir prevents mercury from being drawn into reaction flask or pushed out of bubbler. Do not overfill with mercury. Allows solvents to boil at higher temperatures.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)