Z150819
Worthington LD Series Liquid Nitrogen Dewars
LD5, wide-mouth, capacity 5 L
Synonym(s):
cryo dewar, cryogenic storage, cryopreservation container
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
aluminum flask
Joint
Joints neck (142)
packaging
pack of 1 ea
manufacturer/tradename
Worthington 5LDB
technique(s)
cryopreservation: suitable
capacity
5 L
flask I.D. × depth
19.3 cm (7.6 in.) × 44.5 cm (17.5 in.)
neck I.D.
14.2 cm (5.6 in.)
Looking for similar products? Visit Product Comparison Guide
General description
LD Series Liquid Nitrogen Dewars are designed for storage of small quantities of liquid nitrogen.
High-strength aluminum shell is both durable and light-weight. Super insulation provides advanced thermal protection. Neck tube joins inner and outer container, providing maximum durability and minimum nitrogen consumption. Cap and neck tube core provide positive closure.
High-strength aluminum shell is both durable and light-weight. Super insulation provides advanced thermal protection. Neck tube joins inner and outer container, providing maximum durability and minimum nitrogen consumption. Cap and neck tube core provide positive closure.
Features and Benefits
- Includes a lightweight Snap-On cap for sample protection.
- Superior vacuum performance provides maximum holding times and ultimate thermal protection.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service