Key Documents
Z150681
Sigma-Aldrich
Window for Beckman® Multi-Cell Mount
NaCl, Drilled
About This Item
UNSPSC Code:
12000000
NACRES:
SB.32
material
NaCl
rectangular
description
Polished
technique(s)
IR spectroscopy: suitable
L × W × H
29.5 mm × 14.5 mm × 4 mm
Legal Information
Beckman is a registered trademark of Beckman Coulter, Inc.
Multi-Cell Mount is a trademark of Beckman Coulter, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)