Key Documents
Z147923
Sigma-Aldrich
Glass sample holder
Boat, size 2 1/2 in. × 10 in.
Select a Size
1 EA
$179.00
$179.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$179.00
About This Item
UNSPSC Code:
41100000
NACRES:
NB.43
$179.00
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
borosilicate glass
parameter
230-490 °C temp. limit
size
2 1/2 in. × 10 in.
General description
These sample holders come in both boat and tube designs, made of borosilicate glass. The glass vessel is ideal for transporting and drying chemical samples in the Aldrich Abderhalden apparatus, in desiccators, or in any drying oven. It has a molded end piece to grip and easier handling of the boat.
Choose from one of the most recent versions:
Certificates of Analysis (COA)