Key Documents
Z146552
Ace Rotary-Evaporator/Freeze-Drying Flask Adapter Head W/ O-ring
joint: ST/NS 29/42
Select a Size
1 EA
$573.00
$573.00
Estimated to ship onMarch 30, 2025
Select a Size
Change View
1 EA
$573.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
$573.00
Estimated to ship onMarch 30, 2025
Joint
joint: ST/NS29/42
description
with FETFE®
O-ring.
manufacturer/tradename
Ace Glass 703023
General description
Flange with two ground flats opposite each other allows nylon retainers on adapter head to pass over. O-ring in head and flask grooves make a vacuum-tight seal when head is turned 90° in either direction. No grease, springs, or clamp necessary. Flasks sold separately.
Legal Information
FETFE is a registered trademark of Ace Glass, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)