Key Documents
Z137243
Sigma-Aldrich
Ace three-neck round-bottom flask with threaded side-arm
capacity 500 mL
Synonym(s):
bottom flask, flask round, round flask
Select a Size
1 EA
$285.00
$285.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
Select a Size
Change View
1 EA
$285.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
$285.00
PRICE WITHOUT NATIONAL TAXES
Estimated to ship onJune 02, 2025
material
borosilicate glass
round bottom flask
three-neck (Vertical)
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Ace Glass 6954-75
capacity
500 mL
General description
Three-neck, vertical. 24/40 joints. One threaded side-arm with bushing and O-ring, except 500 mL size which has angled side necks.
Choose from one of the most recent versions:
Certificates of Analysis (COA)