Key Documents
Z136468
Pipette fillers
red natural rubber bulb type
Select a Size
1 EA
$48.60
$48.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
Select a Size
Change View
1 EA
$48.60
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
$48.60
PRICE WITHOUT NATIONAL TAXES
Check Cart for Availability
material
red natural rubber bulb
manufacturer/tradename
RED-001
General description
Valved, choice of natural or silicone rubber bulb. Allows safe manual control of filling and discharging of liquids by finger pressure.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Questions
  1. Does Z136468 pipette filler fit 10 ml (7.87mm diameter) and 1 ml (4.79 mm diameter) pipettes
    1 answer
    1. This natural rubber filler has a 1/4 in. or 6.35 mm diameter opening and will accommodate pipettes up to 50 mL. The 4.79 mm diameter pipette would not be suitable.
      Helpful?