Key Documents
Z129259
Sigma-Aldrich
Aldrich® vacuum-filtration adapter
male joint: ST/NS 29/26, flange I.D. 31 mm
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
Joint
male joint: ST/NS29/26
flange I.D.
31 mm
General description
Enables filtration (via a Buchner funnel) directly into a single-neck, round-bottom flask. Includes rubber sleeve.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)